Would you like to log in again?
rUTP, 100 mM, 400 µl, Promega
Supplier: Promega
E6021
PAE6021EA 94.87 USD InStock
PAE6021
rUTP, 100 mM, 400 µl, Promega
Nucleic Acid Reagents Nucleotides
High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.
  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400µl rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC
Ribonucleotide Triphosphates (rNTPs) are provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty