Would you like to log in again?
ONE-Glo™ Luciferase Assay System, Promega
Supplier: Promega
E6110 E6130 E6120
PAE6110EA 199.92 USD InStock
PAE6110 PAE6120 PAE6130
ONE-Glo™ Luciferase Assay System, Promega
Assays Enzyme Assays
The ONE-Glo Luciferase Assay System provides a sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideal for high- and ultrahigh-throughput applications.
  • Robust Performance for High- and Ultrahigh-Throughput Reporter Assays
  • Less sensitive to mixing and dispensing conditions, enhancing reproducibility
  • Ideal for use in high-density (384- and 1536-well) microplates
  • Reduced odor, and convenient storage options
The ONE-Glo Luciferase Assay System provides a highly sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideally suited for high- and ultrahigh-throughput applications, the ONE-Glo Assay contains a new luciferase substrate, resulting in a reagent that is more stable, more tolerant to sample components, and has less odor than standard luciferase assay reagents. These features ensure that the ONE-Glo Assay provides robust performance and also eliminates many of the handling inconveniences experienced using other reporter assays in a high-throughput setting.
These features ensure that the ONE-Glo Assay provides robust performance and also eliminates many of the handling inconveniences experienced using other reporter assays in a high-throughput setting.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty