Would you like to log in again?
pGL4.18[luc2P/Neo] Vector, 20 µg, Promega
Supplier: Promega
E6731
PAE6731EA 486.08 USD InStock
PAE6731
pGL4.18[luc2P/Neo] Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
The pGL4.17[luc2/Neo] Vector, pGL4.18[luc2P/Neo] and pGL4.19[luc2CP/Neo] Vectors are optimized for expression in mammalian cells. These vectors offer standard and destablized luciferase with neomycin for stable selection.
  • Reporter Vectors That Lack a Promoter and Offer Different Reporter Protein Stabilities and Neomycin Resistance
  • Designed for cloning a putative promoter element for investigation of gene transcription control regions
  • Available with three varieties of engineered firefly luciferase genes: luc2, luc2P or luc2CP
  • Select for stably transfected cells using neomycin (G418)
Promoterless firefly luciferase vectors are designed primarily to accept a putative promoter element for investigation of important regions controlling gene transcription. The promoterless vectors are available with three varieties of engineered firefly luciferase genes: luc2, luc2P or luc2CP. The luc2 gene is engineered to remove most cryptic transcription factor binding sites and improve mammalian expression through codon optimization. The luc2P and luc2CP and RapidResponse genes are luc2 genes appended with degradation sequences to influence the cellular half-life of the luc2 gene. The RapidResponse genes respond more rapidly to stimuli but at the expense of signal intensity. The luc2P (1-hour half-life) gene responds more rapidly than luc2 (3-hour half-life) with moderate signal intensity, and the luc2CP (0.4-hour half-life) responds more quickly with the lowest signal intensity. The promoterlesspGL4.17[luc2/Neo] Vector, pGL4.18[luc2P/Neo] and pGL4.19[luc2CP/Neo] Vectors are available with a selectable marker, neomycin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty