Would you like to log in again?
pGL4.74[hRluc/TK] Vector, 20 µg, Promega
Supplier: Promega
E6921
PAE6921EA 514.26 USD InStock
PAE6921
pGL4.74[hRluc/TK] Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
pGL4.73[hRluc/SV40], pGL4.74[hRluc/TK] and pGL4.75[hRluc/CMV] Vectors are used as the control reporter for dual-luciferase assays. Select the appropriate promoter or test all three in your experimental system.
  • Vectors Co-transfected in Dual-Reporter Assays as the Control Vector
  • Co-transfect into cells where firefly luciferase is the experimental reporter gene
  • CMV, SV40 and TK promoter options offer different expression levels suitable for most dual-reporter assays
The Promoter-driven Renilla control vectors are commonly co-transfected with experimental firefly luciferase vectors for use in the Dual-Luciferase or Dual-Glo Reporter Assay Systems. The control Renilla vectors should give an almost invariant level of activity, while the experimental firefly vector varies with treatment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty