Would you like to log in again?
ONE-Glo + Tox Luciferase Reporter and Cell Viability Assay, Promega
Supplier: Promega
E7110 E7120
PAE7110EA 321.91 USD InStock
PAE7110 PAE7120
ONE-Glo + Tox Luciferase Reporter and Cell Viability Assay, Promega
Assays Cellular Assays
The ONE-Glo + Tox Assay combines luciferase assay chemistry with a cell viability marker to better understand reporter gene expression in the context of cell health. The assay uses a two-step, addition-only process to measure in a single well of a plate.
  • Sensitive Detection of Reporter Activity and Cell Health
  • Measures cell viability and firefly luciferase activity in the same assay well
  • Easy to use—simply add reagent, mix and read
  • Scalable to meet throughput needs, up to 1,536-well format
The ONE-Glo + Tox Assay combines luciferase assay chemistry with a cell viability marker to better understand reporter gene expression in the context of cell health. The assay uses a two-step, addition-only process to make these measurements in a single well of a plate, negating the need to run parallel assays. The first part of the assay is a nonlytic fluorescence assay (CellTiter-Fluor Cell Viability Assay) that measures the relative number of live cells in a culture population after experimental manipulation. The CellTiter-Fluor Assay measures a conserved and constitutive protease activity within live cells and therefore serves as a marker of cell viability. The live-cell protease activity is restricted to intact viable cells and is measured using a fluorogenic, cell-permeant peptide substrate (glycylphenylalanyl-aminofluorocoumarin; GF-AFC). The substrate enters intact cells where it is cleaved by the live-cell protease to generate a fluorescent signal proportional to the number of living cells. This live-cell protease becomes inactive upon loss of cell membrane integrity and leakage into the surrounding culture medium. Fluorescence of the free AFC fluorophore is measured with a microplate reader or CCD imager using an excitation wavelength of 380-400nm and emission wavelength of 505nm. The second part of the assay uses the ONE-Glo Luciferase Assay System to quantify firefly luciferase reporter gene expression from cells made to express this reporter enzyme. The ONE-Glo Luciferase Assay Buffer and ONE-Glo Luciferase Assay Substrate, provided with this system, are combined to form the ONE-Glo Reagent. Ideally suited for high- and ultrahigh-throughput applications, the ONE-Glo Assay contains a new fluoroluciferin substrate, resulting in a more stable reagent that is more tolerant to sample components and has less odor than standard luciferase assay reagents. Luminescence is measured with a microplate reader or CCD imager.
The second part of the assay uses the ONE-Glo Luciferase Assay System to quantify firefly luciferase reporter gene expression from cells made to express this reporter enzyme. The ONE-Glo Luciferase Assay Buffer and ONE-Glo Luciferase Assay Substrate, provided with this system, are combined to form the ONE-Glo Reagent. Ideally suited for high and ultrahigh-throughput applications, the ONE-Glo Assay contains a new fluoroluciferin substrate, resulting in a more stable reagent that is more tolerant to sample components and has less odor than standard luciferase assay reagents. Luminescence is measured with a microplate reader or CCD imager.
The first part of the assay is a nonlytic fluorescence assay (CellTiter-Fluor™ Cell Viability Assay) that measures the relative number of live cells in a culture population after experimental manipulation. The CellTiter-Fluor Assay measures a conserved and constitutive protease activity within live cells and therefore serves as a marker of cell viability. The live-cell protease activity is restricted to intact viable cells and is measured using a fluorogenic, cell-permeant peptide substrate (glycylphenylalanyl-aminofluorocoumarin, GF-AFC). The substrate enters intact cells where it's cleaved by the live-cell protease to generate a fluorescent signal proportional to the number of living cells. This live-cell protease becomes inactive upon loss of cell membrane integrity and leakage into the surrounding culture medium. Fluorescence of the free AFC fluorophore is measured with a microplate reader or CCD imager with an excitation wavelength of 380–400 nm and emission wavelength of 505 nm.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty