Would you like to log in again?
ONE-Glo EX Luciferase Assay System, Promega
Supplier: Promega
E8110 E8130 E8150 E8120
PAE8110EA 200.57 USD InStock
PAE8110 PAE8150 PAE8130 PAE8120
ONE-Glo EX Luciferase Assay System, Promega
Assays Enzyme Assays
ONE-Glo EX Luciferase Assay System provides both the high sensitivity and long-lived luminescence required to batch-process multiple 96-, 384- or 1,536-well plates. The approximately 2-hour signal half-life provides greater flexibility in assay design.
  • Highly Sensitive Assay with Long-Lived Luminescence
  • Bright, stable signal that can be measured for hours
  • Stable reagent can be stored at room temperature or 4°C
  • Reduced sensitivity to culture media, phenol red and luciferase inhibitors
High- or ultrahigh-throughput quantitation of firefly luciferase expression in mammalian cells is commonly performed by measuring luminescence from 96-, 384- or 1,536-well plates. The ONE-Glo EX Luciferase Assay System provides both the high sensitivity and long-lived luminescence required to batch-process multiple plates in these assay formats. The ONE-Glo EX Assay retains many of the beneficial aspects of the ONE-Glo Assay, using 5´-fluoroluciferin substrate with an add-mix-read, or homogeneous, protocol. Extending the properties of ONE-Glo Reagent, ONE-Glo EX Reagent employs a new assay chemistry to increase the stability of both the luminescence signal and the reconstituted reagent. The approximately 2-hour signal half-life provides greater flexibility in assay design. A reconstituted reagent that can be stored at room temperature for longer periods means less variability in reagent performance during long experiments or screens and less sample waste. ONE-Glo EX Reagent is the firefly luciferase detection reagent used in the Nano-Glo Dual-Luciferase Reporter (NanoDLR) Assay System, allowing the same reagent to be used for single- or dual-luciferase assays.
The approximately two-hour signal half life provides greater flexibility in assay design. A reconstituted reagent that can be stored at room temperature for longer periods means less variability in reagent performance during long experiments or screens and less sample waste. ONE-Glo EX Reagent is the firefly luciferase detection reagent used in the Nano-Glo® Dual-Luciferase® Reporter (NanoDLR™) Assay System, allowing the same reagent to be used for single or dual-luciferase assays.
The ONE-Glo EX Assay retains many of the beneficial aspects of the ONE-Glo Assay, using 5´-fluoroluciferin substrate with an add-mix-read, or homogeneous, protocol. Extending the properties of ONE-Glo Reagent, ONE-Glo EX Reagent employs a new assay chemistry to increase the stability of both the luminescence signal and the reconstituted reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty