Would you like to log in again?
ENLITEN rLuciferase/Luciferin Reagent, Promega
Supplier: Promega
ENLITEN®
FF2021
PAFF2021EA 254.61 USD InStock
PAFF2021
ENLITEN rLuciferase/Luciferin Reagent, Promega
Assays Enzyme Assays
The ENLITEN rLuciferase/Luciferin Reagent is intended for the rapid and quantitative detection of ATP in liquid samples.
  • Detect and Quantitate ATP in Liquid Samples
The ENLITEN rLuciferase/Luciferin Reagent is intended for the rapid and quantitative detection of ATP in liquid samples. The reagent is designed to measure 10(-11) to 10(-15) moles of ATP. Some of the applications may include the indirect measurement of bacteria, yeasts and fungi on surfaces or in products, assaying enzymes that degrade ATP or quantitation of ATP in biological fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty