Would you like to log in again?
CellTiter 96 AQueous MTS Reagent Powder, Promega
Supplier: Promega
G1112 G1111
PAG1112EA 465.27 USD InStock
PAG1112 PAG1111
CellTiter 96 AQueous MTS Reagent Powder, Promega
Assays Cellular Assays
CellTiter 96(R) AQueous MTS Reagent Powder is a novel tetrazolium compound for use in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays.
  • A Novel Tetrazolium Compound for Use in Colorimetric Assays
  • Available in two sizes
  • Use in assays to determine the number of viable cells
CellTiter 96(R) AQueous MTS Reagent Powder is a novel tetrazolium compound for use in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays. It can be purchased separately and is provided in powdered form.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty