Would you like to log in again?
Lambda DNA/HindIII Markers, 100 µg, Promega
Supplier: Promega
G1711
PAG1711EA 78.73 USD InStock
PAG1711
Lambda DNA/HindIII Markers, 100 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Generated by digesting lambda DNA to completion with HindIII, resulting in DNA fragments of 125-23,130bp.
  • Eight DNA Fragments of 125-23,130bp
  • Load 1microl/lane
  • Supplied at a concentration of 0.5µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. Lambda DNA/HindIII Markers: Eight ethanol-precipitated DNA fragments ranging in size from 125bp to 23,130bp. Recommended loading: 1microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty