Would you like to log in again?
CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays, Promega
Supplier: Promega
G1780
PAG1780EA 439.29 USD InStock
PAG1780
CytoTox 96 Non-Radioactive Cytotoxicity Assay, 1,000 assays, Promega
Assays Cellular Assays
Colorimetric assay for the determination of cell membrane integrity based on detection of LDH release, an indicator of cytotoxicity. Sample transfer steps are required.
  • A Colorimetric Alternative to Radioactive Cytotoxicity Assays
  • Measures release of LDH, an indicator of cytotoxicity
  • Non-radioactive, time-saving assay
The CytoTox 96 Non-Radioactive Cytotoxicity Assay is a colorimetric alternative to radioactive cytotoxicity assays. The CytoTox 96 Assay quantitatively measures lactate dehydrogenase (LDH), a stable cytosolic enzyme that is released upon cell lysis, in much the same way as [(51)Cr] is released in radioactive assays. Released LDH in culture supernatants is measured with a 30-minute coupled enzymatic assay that results in the conversion of a tetrazolium salt (INT) into a red formazan product. The amount of color formed is proportional to the number of lysed cells. Visible wavelength absorbance data are collected using a standard 96-well plate reader. The assay can be used to measure membrane integrity for cell-mediated cytotoxicity assays in which a target cell is lysed by an effector cell, or to measure lysis of target cells by bacteria, viruses, proteins, chemicals, etc.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty