Would you like to log in again?
DNA Step Ladder 50 bp, 90 µg, Promega
Supplier: Promega
G4521
PAG4521EA 223.29 USD InStock
PAG4521
DNA Step Ladder 50 bp, 90 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Sixteen DNA fragments ranging from 50bp to 800bp in 50bp increments plus an 1,800bp "backbone" fragment.
  • Sixteen DNA Fragments of 50-800bp in 50bp Increments
  • Includes an 1,800bp "backbone" fragment
  • Load 5microl/lane
  • Supplied at a concentration of 0.34µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 50bp DNA Step Ladder: Sixteen DNA fragments ranging from 50bp to 800bp in 50bp increments plus an 1,800bp backbone fragment. All bands except the 800bp band are of equal intensity; the 800bp band is ~3 times more intense. Recommended loading: 5microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty