Would you like to log in again?
CellTiter 96® AQueous Non-Radioactive Cell Proliferation Assay, Promega
Supplier: Promega
G5421 G5430 G5440
PAG5421EA 355.75 USD InStock
PAG5421 PAG5430 PAG5440
CellTiter 96® AQueous Non-Radioactive Cell Proliferation Assay, Promega
Assays Cellular Assays
Homogeneous, colorimetric cell viability assay; requires mixing of MTS and PMS components prior to assay, then a 1- to 4-hour incubation. Standalone MTS powder for applications where optimization of assay for specific cell types or conditions is desired.
  • A Colorimetric Method for Determining the Number of Viable Cells
  • Measures viable cells in proliferation, cytotoxicity or chemosensitivity assays
  • Non-radioactive assay makes it safe and easy use
The CellTiter 96 AQueous Non-Radioactive Cell Proliferation Assay is a homogeneous, colorimetric method for determining the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays. The CellTiter 96 AQueous Assay is composed of solutions of a novel tetrazolium compound [3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium, inner salt; MTS] and an electron coupling reagent (phenazine methosulfate) PMS. MTS is bioreduced by cells into a formazan product that is soluble in tissue culture medium. The absorbance of the formazan product at 490nm can be measured directly from 96-well assay plates without additional processing. The conversion of MTS into the aqueous soluble formazan product is accomplished by dehydrogenase enzymes found in metabolically active cells. The quantity of formazan product as measured by the amount of 490nm absorbance is directly proportional to the number of living cells in culture. If you currently use a [(3)H]-thymidine incorporation assay, addition of the combined MTS/PMS solution can be substituted for [(3)H]-thymidine at the time point in the assay when the pulse of radioactive thymidine is usually added. Data from proliferation bioassays comparing the CellTiter 96 AQueous Assay and [(3)H]-thymidine incorporation show similar results. This is in agreement with similar radioactivity incorporation studies performed using the original CellTiter 96 Assay. CellTiter 96 AQueous MTS Reagent Powder is a novel tetrazolium compound for use in colorimetric assays for determining the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays. It is provided in powdered form.
If you currently use a [3H]-thymidine incorporation assay, the addition of the combined MTS/PMS solution can be substituted for [3H]-thymidine at the point in the assay when the pulse of radioactive thymidine is usually added. Data from proliferation bioassays comparing the CellTiter 96 AQueous Assay and [3H]-thymidine incorporation show similar results. This is in agreement with similar radioactivity incorporation studies performed using the original CellTiter 96 Assay.
MTS is bioreduced by cells into a formazan product that is soluble in tissue culture medium. The absorption of the formazan product at 490 nm can be measured directly from 96-well assay plates without additional processing. The conversion of MTS into the aqueous soluble formazan product is accomplished by dehydrogenase enzymes found in metabolically active cells. The quantity of formazan product as measured by the amount of 490 nm absorbance is directly proportional to the number of living cells in culture.
Accessories information: CellTiter 96 AQueous MTS Reagent Powder is available separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty