Would you like to log in again?
CellTiter-Fluor Cell Viability Assay, Promega
Supplier: Promega
G6082 G6080 G6081
PAG6082EA 1115.01 USD InStock
PAG6082 PAG6081 PAG6080
CellTiter-Fluor Cell Viability Assay, Promega
Assays Cellular Assays
The CellTiter-Fluor Cell Viability Assay is a non-lytic, single-reagent-addition fluorescence assay that measures the relative number of viable cells in a population.
  • A Non-Lytic Fluorescence Assay That Measures the Relative Number of Viable Cells in Culture
  • Multiplex with many other luminescent assays
  • Normalizing data for live-cell numbers make results more comparable to other data
The CellTiter-Fluor Cell Viability Assay is a non-lytic, single-reagent-addition fluorescence assay that measures the relative number of viable cells in a population. The assay is based on measurement of a conserved and constitutive protease activity within live cells and therefore serves as a biomarker of cell viability. The live-cell protease activity is restricted to intact viable cells and is measured using a fluorogenic, cell-permeant, peptide substrate (Gly-Phe-AFC). The substrate enters intact cells, where it is cleaved by the live-cell protease activity to generate a fluorescent signal proportional to the number of living cells. The live-cell protease becomes inactive upon loss of membrane integrity and leakage into the surrounding culture medium. The CellTiter-Fluor Assay also can be used in a single-well, sequential, multiplex format with other downstream assay chemistries to normalize data by cell number. Data from the assay can serve as an internal control and allow identification of errors resulting from cell clumping or compound cytotoxicity. The assay is compatible with many Promega luminescence assays or spectrally distinct fluorescence assay methods, such as measuring caspase activation, reporter gene expression or orthogonal measures of viability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty