Would you like to log in again?
HaloTag Protein Purification System, 1 each, Promega
Supplier: Promega
G6280
PAG6280EA 941 USD InStock
PAG6280
HaloTag Protein Purification System, 1 each, Promega
Protein Purification Protein Purification Resins
HaloTag Protein Purification System is designed to purify proteins fused to the HaloTag novel protein tag that enhances expression and solubility of proteins from E. coli and enables the covalent, efficient and specific capture of a protein of interest.
  • Enhanced Expression and Solubility of Recombinant Proteins with HaloTag(R) Ligands
  • Covalent, efficient and specific capture of your protein of interest
  • Overcome limitations (i.e., poor capture of proteins expressed at low levels, protein loss during washing) associated with affinity tags
  • Sample size is available (choose Cat.# G6270)
The HaloTag Protein Purification System is designed to purify proteins fused to the HaloTag novel protein tag that enhances the expression and solubility of recombinant proteins. HaloTag Technology enables the covalent, efficient and specific capture of a protein of interest onto HaloLink Resin, thus overcoming the equilibrium-based limitations associated with affinity tags (i.e., poor capture of proteins expressed at low levels and protein loss during washing of the purification resin. The HaloTag technology offers a quick and convenient way to test protein expression of HaloTag fusion proteins as well as monitor the efficiency of immobilization to HaloLink Resin by labeling with fluorescent HaloTag TMR Ligand followed by SDS-PAGE analysis. Guidelines can be found in the HaloLink Resin Technical Manual #TM250, the HaloLink Protein Array Technical Manual #TM310 and the HaloCHIP System Technical Manual #TM075. Outline of Procedure: The HaloTag protein, a 34kDa mutated hydrolase, covalently attaches to HaloLink Resin via an immobilized chloroalkane ligand. TEV Protease cleaves the target protein from the HaloLink Resin. The TEV Protease, which has an N-terminal (HQ) tag, is removed from the protein of interest using HisLink Resin, and the purified protein of interest is recovered. The appropriate vector that encodes the HaloTag protein and expresses protein optimally in E. coli is pFN18A HaloTag T7 Flexi Vector (G2751) or pFN18K HaloTag T7 Flexi Vector (G2681). These vectors can be purchased separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty