Would you like to log in again?
Magne Protein G Beads, 20% Slurry, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
G7473 G7471 G7472
PAG7473EA 4397.41 USD InStock
PAG7473 PAG7472 PAG7471
Magne Protein G Beads, 20% Slurry, Promega
Beads Magnetic Beads
Magne Protein G & Magne Protein A Beads are magnetic affinity beads with high specificity and capacity for purification of immunoglobulins from cell culture media, ascites and serum. Iron encapsulated in macroporous cellulose with low nonspecific binding.
  • High Specificity, High Capacity Magnetic Affinity Beads for Antibody Capture
  • Binding capacities of >25 mg/ml depending on antibody species, isotype
  • Minimize purification losses and increase sample throughput with exceptional magnetic properties
Magne Protein G and Magne Protein A Beads are magnetic affinity beads with high specificity and high capacity for purification of immunoglobulins from cell culture media, ascites and serum samples. These paramagnetic beads are composed of iron encapsulated in macroporous cellulose with low nonspecific binding. The magnetic beads use a novel attachment chemistry to immobilize recombinant Protein G or Protein A protein molecules in the same orientation on the surface of the bead. The oriented attachment is known to improve the functionality of immobilized proteins. These beads offer a convenient method for achieving high purity and high recovery of monoclonal and polyclonal antibodies from a variety of biological samples. The superb magnetic properties of Magne Protein G and Magne Protein A Beads allow rapid and efficient capture of antibodies either with manually processed samples or in a high-throughput manner using the Promega ReliaPrep LV 32 HSM Instrument or a robotic platform such as the Beckman Coulter Biomek FX.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty