Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BenchTop PCR Markers, 300 µl, Promega
Supplier: Promega
G7531
PAG7531EA 247.82 USD InStock
PAG7531
BenchTop PCR Markers, 300 µl, Promega
Electrophoresis Reagents Molecular Weight Markers
50-1,000bp; 6 bands. Can be stored at room temperature. Contains dye marker, ready to load on agarose and polyacrylamide gels.
  • Six Bands of Equal Intensity: 50, 150, 300, 500, 750, and 1,000bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 6microl/lane for agarose gels
The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. Six bands of equal intensity of 50, 150, 300, 500, 750, and 1,000bp. The BenchTop PCR Markers may be run on polyacrylamide gels with less loading volume; however, additional bands may be visible compared to those visible on agarose gels. Load 6microl/lane for agarose gels.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty