Would you like to log in again?
Bio-Glo™ Luciferase Assay System, 100 ml, Promega
Supplier: Promega
G7941 G7940
PAG7941EA 271.15 USD InStock
PAG7941 PAG7940
Bio-Glo™ Luciferase Assay System, 100 ml, Promega
Assays Cellular Assays
The Bio-Glo Luciferase Assay System provides a highly sensitive, robust, homogeneous reagent, the Bio-Glo Assay reagent with a new luciferase substrate, for the detection of firefly luciferase reporter gene expression in the ADCC Reporter Bioassay.
  • Stable, Inhibitor-Resistant Luciferase Reagent for Bioassays
  • Brighter, longer-lasting signal enables long incubations
  • Room-temperature storage provides added convenience
The Bio-Glo Luciferase Assay System provides a highly sensitive, robust, homogeneous reagent for the detection of firefly luciferase reporter gene expression in the ADCC Reporter Bioassay. Bio-Glo Assay reagent contains a new luciferase substrate, resulting in a reagent that is more stable and more tolerant to sample components than standard luciferase assay reagents. Bio-Glo Assay reagent is functionally tested for performance in the ADCC Reporter Bioassay and is intended for use with this or other bioassays.
Bio-Glo Assay reagent is functionally tested for performance in the ADCC Reporter Bioassay and is intended for use with this or other bioassays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty