Would you like to log in again?
Mitochondrial ToxGlo Assay, Promega
Supplier: Promega
G8001 G8000
PAG8000EA 306.57 USD InStock
PAG8000 PAG8001
Mitochondrial ToxGlo Assay, Promega
Assays Cellular Assays
The Mitochondrial ToxGlo Assay provides a method for predicting potential mitochondrial dysfunction as a result of xenobiotic exposure.
  • A Cell-Based Method for Predicting Potential Mitochondrial Dysfunction
  • Distinguish primary mitochondrial dysfunction from secondary cytotoxic events
  • Predictive for mitochondrial toxicities
  • Fast, cost-effective and easy to implement
The Mitochondrial ToxGlo Assay is a cell-based assay method that employs a sequential addition, multiplexed assay chemistry for predicting potential mitochondrial dysfunction as a result of xenobiotic exposure. The assay is based on the differential measurement of biomarkers associated with changes in cell membrane integrity and cellular ATP levels relative to vehicle-treated control cells during short exposure periods. Cell membrane integrity is first assessed by measuring the presence or absence of a distinct protease activity associated with necrosis using a fluorogenic peptide substrate (bis-AAF-R110) to measure "dead cell protease activity". The bis-AAF-R110 Substrate cannot cross the intact membrane of live cells and therefore gives no signal with viable cells. Next, ATP is measured by adding an ATP detection reagent, resulting in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The two sets of data can be combined to produce profiles representative of mitochondrial dysfunction or non-mitochondrial related cytotoxic mechanisms.Mammalian cells generate ATP by mitochondrial (oxidative phosphorylation) and non-mitochondrial (glycolysis) methods. To achieve optimal mitochondrial responsiveness, it may be necessary to refine cell culture conditions. Replacing glucose-supplemented medium with galactose-containing medium may increase cellular oxygen consumption and augment mitochondrial susceptibility to mitotoxicants.
Mammalian cells generate ATP by mitochondrial (oxidative phosphorylation) and non-mitochondrial (glycolysis) methods. It may be necessary to refine cell culture conditions to achieve optimal mitochondrial responsiveness. Replacing glucose-supplemented medium with galactose-containing medium may increase cellular oxygen consumption and augment mitochondrial susceptibility to mitotoxicants.
Cell membrane integrity is first assessed by measuring the presence or absence of a distinct protease activity associated with necrosis using a fluorogenic peptide substrate (bis-AAF-R110) to measure dead cell protease activity. The bis-AAF-R110 Substrate cannot cross the intact membrane of live cells and therefore gives no signal with viable cells. Next, ATP is measured by adding an ATP detection reagent, resulting in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The two sets of data can be combined to produce profiles representative of mitochondrial dysfunction or non-mitochondrial related cytotoxic mechanisms.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty