Would you like to log in again?
CellTiter-Blue Cell Viability Assay, Promega
Supplier: Promega
G8082 G8080 G8081
PAG8080EA 162.62 USD InStock
PAG8080 PAG8081 PAG8082
CellTiter-Blue Cell Viability Assay, Promega
Assays Cellular Assays
Homogeneous, fluorometric cell viability assay (resazurin). Requires a 1- to 4-hour incubation.
  • A Homogeneous, Fluorescent Assay to Monitor Cell Viability
  • Fast, flexible, time-saving
  • Amenable to multiplexing with other assays
The CellTiter-Blue Cell Viability Assay provides a homogeneous, fluorescent method for monitoring cell viability. The assay is based on the ability of living cells to convert a redox dye (resazurin) into a fluorescent end product (resorufin). Nonviable cells rapidly lose metabolic capacity and thus do not generate a fluorescent signal. The homogeneous assay procedure involves adding the single reagent directly to cells cultured in serum-supplemented medium. After an incubation step, data are recorded using either a plate-reading fluorometer (preferred) or spectrophotometer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty