Would you like to log in again?
Caspase-Glo 3/7 Assay Systems, Promega
Supplier: Promega
G8093 G8090 G8091 G8092
PAG8090EA 168.34 USD InStock
PAG8090 PAG8091 PAG8092 PAG8093
Caspase-Glo 3/7 Assay Systems, Promega
Assays Cellular Assays
Caspase-Glo 3/7 Assay is a homogeneous luminescent assay that measures caspase-3/7 activities
  • A Luminescent Assay to Measure Caspase-3/7 Activities Using Fewer Cells
  • Superior sensitivity and linearity over four orders of magnitude of concentration
  • Homogeneous and highly reproducible assay
  • Uses less enzyme and no sample preparation required
The Caspase-Glo 3/7 Assay provides a homogeneous luminescent assay that measures caspase-3/7 activities. The assay provides a proluminescent caspase-3/7 DEVD-aminoluciferin substrate and a proprietary thermostable luciferase in a reagent optimized for caspase-3/7 activity, luciferase activity and cell lysis. Adding the single Caspase-Glo 3/7 Reagent in an add-mix-measure format results in cell lysis, followed by caspase cleavage of the substrate. This liberates free aminoluciferin, which is consumed by the luciferase, generating a glow-type luminescent signal. The signal is proportional to caspase-3/7 activity. The stabilized luciferase and proprietary buffer system improve assay performance across a wide range of assay conditions, and the assay is less likely to be affected by compound interference unlike fluorescent- or colorimetric-based assays. The Caspase-Glo 3/7 Assay is designed for use with multiwell plate formats using either purified enzyme or cells in culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty