Would you like to log in again?
Caspase-Glo 8 Assay, Promega
Supplier: Promega
G8201 G8200 G8202
PAG8200EA 173.36 USD InStock
PAG8200 PAG8201 PAG8202
Caspase-Glo 8 Assay, Promega
Assays Cellular Assays
Caspase-Glo 8 Assay is a homogeneous luminescent assay that measures caspase-8 activity.
  • A Luminescent Assay That Measures Caspase-8 Activity
  • Decreased assay time and less enzyme needed
  • Homogeneous “add-mix-measure” protocol amenable to automation
The Caspase-Glo 8 Assay is a homogeneous luminescent assay that measures caspase-8 activity. The assay provides a proluminogenic caspase-8 substrate in a buffer system optimized for caspase activity, luciferase activity and cell lysis. The addition of a single Caspase-Glo 8 Reagent in an add-mix-read format results in cell lysis, followed by caspase cleavage of the substrate and generation of a glow-type luminescent signal. The signal generated is proportional to the amount of caspase activity present. The Caspase-Glo Reagent relies on the properties of a proprietary thermostable luciferase (Ultra-Glo Recombinant Luciferase), which generates the stable glow-type luminescent signal and improves performance across a wide range of assay conditions. The system now includes a separate vial of a protease inhibitor, MG-132 Inhibitor, which may be used to reduce background, thus improving the performance of the Caspase-Glo 8 Assay in cell-based applications.
The system now includes a separate vial of a protease inhibitor, MG-132 Inhibitor, which may be used to reduce background, thus improving the performance of the Caspase-Glo 8 Assay in cell-based applications.
The addition of a single Caspase-Glo 8 Reagent in an add-mix-read format results in cell lysis, followed by caspase cleavage of the substrate and generation of a glow-type luminescent signal. The signal generated is proportional to the amount of caspase activity present. The Caspase-Glo Reagent relies on the properties of a proprietary thermostable luciferase (Ultra-Glo™ Recombinant Luciferase), which generates the stable glow-type luminescent signal and improves performance across a wide range of assay conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty