Would you like to log in again?
BacTiter-Glo™ Microbial Cell Viability Assay, Promega
Supplier: Promega
BacTiter-Glo™
G8233 G8230 G8232 G8231
PAG8233EA 5121.83 USD InStock
PAG8233 PAG8230 PAG8231 PAG8232
BacTiter-Glo™ Microbial Cell Viability Assay, Promega
Assays Cellular Assays
The BacTiter-Glo Microbial Cell Viability Assay is a homogeneous method for determining the number of viable microbial cells in culture based on quantitation of the ATP present.
  • A Luminescent Assay to Measure the Number of Viable Microbial Cells in Culture
  • A homogeneous, single reagent assay
  • Offers increased sensitivity, fast results
The BacTiter-Glo Microbial Cell Viability Assay is a homogeneous method for determining the number of viable microbial cells in culture based on quantitation of the ATP present. ATP is an indicator of metabolically active cells. The homogeneous assay procedure involves adding a single reagent (BacTiter-Glo Reagent) directly to bacterial cells cultured in medium and measuring luminescence. The homogeneous format reduces pipetting errors that may be introduced during the multiple steps required by other methods of ATP measurement. The formulation of the reagent supports bacterial cell lysis and generation of a luminescent signal in a homogeneous add-mix-measure format. The luminescent signal is proportional to the amount of ATP present, which is directly proportional to the number of viable cells in culture. The assay relies on the properties of a proprietary thermostable luciferase (Ultra-Glo Recombinant Luciferase) and a proprietary buffer formulation for extracting ATP from bacteria. The assay has been shown to detect a variety of bacteria and fungi.
Data can be recorded in as little as five minutes with a superior sensitivity that allows growth or toxicity detection to quickly follow inoculation. Users can draw ATP measurements from as few as 10 bacterial cells. The assay can be used with various multiwell plates or single-use formats. Data can be recorded by luminometer or CCD camera. Store at –20°C (–4°F).
The luminescent signal is proportional to the amount of ATP present, which is directly proportional to the number of viable cells in culture. This add-mix-measure format reduces the number of handling steps and the potential for procedural error. The procedure has no separate lysis step, and no injectors are required, allowing easy automation.
Ordering information: Each assay is available in four different sizes and includes BacTiter-Glo buffer, substrate (lyophilized), and one protocol.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty