Would you like to log in again?
DPPIV-Glo Protease Assay, 10ml, Promega
Supplier: Promega
G8350 G8351
PAG8350EA 491.16 USD InStock
PAG8350 PAG8351
DPPIV-Glo Protease Assay, 10ml, Promega
Assays Cellular Assays
The DPPIV-Glo Protease Assay is a homogeneous, luminescent assay that measures dipeptidyl peptidase IV (DPPIV) activity.
  • Homogeneous, Luminescent Assay that Measures Dipeptidyl Peptidase IV Activity
  • Simple "add-mix-measure" protocol
  • Results in as little as 30 minutes after reagent addition
  • Signal stability allows batch processing
The DPPIV-Glo Protease Assay is a homogeneous, luminescent assay that measures dipeptidyl peptidase IV (DPPIV) activity. DPPIV is a serine protease that cleaves N-terminal dipeptides from polypeptides with L-proline or L-alanine at the penultimate position. The DPPIV-Glo Assay provides a proluminescent DPPIV substrate, Gly-Pro-aminoluciferin, in a buffer system optimized for DPPIV and luciferase activities. The addition of a single DPPIV-Glo Reagent in an add-mix-measure format results in DPPIV cleavage of the substrate and generation of a glow-type luminescent signal produced by the luciferase reaction. In this homogeneous, coupled-enzyme format, the signal is proportional to the amount of DPPIV activity present. The assay is designed for use with purified enzyme preparations.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty