Would you like to log in again?
CellTiter-Glo One Solution Assay, Promega
Supplier: Promega
G8462 G8461
PAG8461EA 565.78 USD InStock
PAG8461 PAG8462
CellTiter-Glo One Solution Assay, Promega
Assays Cellular Assays
The CellTiter-Glo One Solution Assay is a homogeneous method of determining the number of viable cells in culture based on quantitation of the ATP present, which indicates the presence of metabolically active cells.
  • Luminescent Assay Measures Number of Viable Cells in Culture
  • Frozen, ready-to-use assay eliminates several protocol steps
The CellTiter-Glo One Solution Assay is a homogeneous method of determining the number of viable cells in culture based on quantitation of the ATP present, which indicates the presence of metabolically active cells. The frozen, ready-to-use format is based on the original CellTiter-Glo Luminescence Cell Viability Assay chemistry and eliminates the need to combine buffer with lyophilized substrate when preparing reagent. The CellTiter-Glo Assay is designed for use with multiwell-plate formats, making it ideal for automated high-throughput screening (HTS) in 96- to 1536-well format, and cell proliferation and cytotoxicity assays. The homogeneous "add-mix-measure" format results in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The amount of ATP is directly proportional to the number of cells present in culture. The CellTiter-Glo One Solution Assay generates a stable "glow-type" luminescent signal with a half-life of greater than three hours. This extended half-life eliminates the need for reagent injectors and provides flexibility for continuous or batch-mode processing of multiple plates.
The homogeneous add-mix-measure format results in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The amount of ATP is directly proportional to the number of cells present in culture. The CellTiter-Glo One Solution Assay generates a stable glow-type luminescent signal with a half-life of greater than three hours. This extended half-life eliminates the need for reagent injectors and provides flexibility for continuous or batch-mode processing of multiple plates. 100 mL of One Solution Assay is sufficient for 1000 assays at 100 µL/assay in 96-plates or 4000 assays at 25 µL/assay in 384-well plates. 500 mL of One Solution Assay is sufficient for 5000 assays at 100 µL/assay or 20,000 assays at 25 µL/assay in 384-well plates.
The CellTiter-Glo Assay is designed for use with multiwell-plate formats, making it ideal for automated high-throughput screening (HTS) in 96 to 1536-well format and cell proliferation and cytotoxicity assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty