Would you like to log in again?
Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2×50 ml, Promega
Supplier: Promega
G8662
PAG8662EA 3519.53 USD InStock
PAG8662
Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2×50 ml, Promega
Assays Cellular Assays
The Proteasome-Glo Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.
  • Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
  • Simple "add-mix-measure" protocol
  • Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
  • Results within 10-30 minutes after reagent addition
The Proteasome-Glo Cell-Based Assays are homogeneous, luminescent assays that individually measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells. The 26S proteasome is a 2.5MDa multiprotein complex found in all eukaryotic cells. Proteasome-Glo Cell-Based Assays provide luminogenic proteasome substrates in buffers optimized for cell permeabilization, proteasome activity and luciferase activity. Addition of the Proteasome-Glo Cell-Based Reagent in an add-mix-measure format results in proteasome cleavage of the substrate and rapid generation of a luminescent signal produced by the luciferase reaction. The three luminogenic substrates used to monitor specific protease activities include: Suc-LLVY-aminoluciferin for chymotrypsin-like, Z-LRR-aminoluciferin for trypsin-like, and Z-nLPnLD-aminoluciferin for caspase-like activity. Each luminogenic substrate is added to a buffer system optimized for its specific proteasome activity and luciferase activity. The reagents are added to cells in culture, and the proteasome cleaves the substrates, releasing luciferin, which is consumed by luciferase, producing glow-type luminescence correlating to enzyme activity or inhibition. The Proteasome-Glo Cell-Based 3-Substrate System consists of three homogeneous bioluminescent assays that measure the three proteolytic activities associated with the proteasome in a cell-based format (each of these three assays also is available separately). The Proteasome-Glo 3-Substrate System consists of three homogeneous bioluminescent assays in a purified enzyme-based format (each of these three assays also is available separately).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty