Would you like to log in again?
ROS-Glo H2O2 Assay, Promega
Supplier: Promega
G8820 G8821
PAG8820EA 516.09 USD InStock
PAG8820 PAG8821
ROS-Glo H2O2 Assay, Promega
Assays Enzyme Assays
The ROS-Glo H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions.
  • Specific, Direct Detection of Hydrogen Peroxide (H2O2)
  • Fast assay involves a simple two-reagent-addition protocol
  • Substrate reacts directly with H2O2; no need for HRP as a coupling enzyme
The ROS-Glo H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions. A derivatized luciferin substrate is incubated with sample and reacts directly with H2O2 to generate a luciferin precursor. Addition of ROS-Glo Detection Solution converts the precursor to luciferin and provides Ultra-Glo Recombinant Luciferase to produce light signal that is proportional to the level of H2O2 present in the sample.
The addition of ROS-Glo Detection Solution converts the precursor to luciferin and provides Ultra-Glo™ Recombinant Luciferase to produce light signal that is proportional to the level of H2O2 present in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty