Would you like to log in again?
CytoTox-Fluor Cytotoxicity Assay, 10 ml, Promega
Supplier: Promega
G9260 G9262 G9261
PAG9260EA 168.7 USD InStock
PAG9260 PAG9261 PAG9262
CytoTox-Fluor Cytotoxicity Assay, 10 ml, Promega
Assays Cellular Assays
The CytoTox-Fluor Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the relative number of dead cells in cell populations.
  • A Fluorescent Assay to Measure the Relative Number of Dead Cells in Culture
  • Single-reagent-addition, homogeneous assay
  • Amenable to multiplexing with other luminescent cell-based assays
The CytoTox-Fluor Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the relative number of dead cells in cell populations. The assay measures a distinct protease activity associated with cytotoxicity and uses a fluorogenic peptide substrate (bis-alanyl-alanyl-phenylanlanyl-rhodamine 110; bis-AAF-R110) to measure dead-cell activity, which has been released from cells that have lost membrane integrity. The bis-AAF-R110 substrate cannot cross the intact membrane of live cells and therefore gives no signal from live cells. The assay is designed to accommodate downstream multiplexing with several Promega luminescent assays or spectrally distinct fluorescent assay methods, such as assays to measure caspase activation, reporter gene expression or orthogonal measures of viability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty