Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GloMax Explorer Fully Loaded Model, Promega
Supplier: Promega
GM3500
PAGM3500EA 31893.1 USD InStock
PAGM3500
GloMax Explorer Fully Loaded Model, Promega
Microplate Readers
The GloMax Explorer System is a high-performance multimode detection instrument developed with Promega reagents to provide a simple means of detecting advanced chemistries.
  • Multimode Reader with Maximum Flexibility
  • Fully loaded model detects luminescence, fluorescence and absorbance
  • Integrated tablet PC provides quick and easy navigation
  • Developed and optimized with our cell and gene reporter assays
The GloMax Explorer System is a high-performance multimode detection instrument developed with Promega reagents to provide a simple means of detecting advanced chemistries. This instrument provides researchers superior luminescence sensitivity and dynamic range for both strong and weak bioluminescence experimental samples as well as seamless integration with Promega industry-leading bioluminescence assays. GloMax Explorer provides flexible use of filters for fluorescence intensity, filtered luminescence and visible absorbance measurements. The instrument is operated by an integrated tablet PC, which provides quick and easy navigation through the control options. Exporting your results is easy with a variety of options, including export to your local data network. The GloMax Explorer software provides many of the required technical elements of a part 11-compliant system (user authentication and authorization, data integrity and protection, electronic signatures and audit trails) when used with the appropriate laboratory workflow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty