Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GloMax Explorer Absorbance Module Upgrade, Promega
Supplier: Promega
GM3520
PAGM3520EA 8392.79 USD InStock
PAGM3520
GloMax Explorer Absorbance Module Upgrade, Promega
Microplate Readers
These accessories are intended for use with the GloMax Discover and Explorer Systems.
  • For use with GloMax(R) Discover and Explorer Systems
  • Filter Paddles only available for GloMax(R) Discover System
  • Absorbance Module only available for GloMax(R) Explorer System
These accessories are intended for use with the GloMax Discover and Explorer Systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty