Would you like to log in again?
S30 T7 High-Yield Protein Expression System, Promega
Supplier: Promega
L1110 L1115
PAL1110EA 692.37 USD InStock
PAL1110 PAL1115
S30 T7 High-Yield Protein Expression System, Promega
Protein Expression Protein Expression Systems
The E. coli S30 T7 High-Yield Protein Expression System is designed to express up to 500ug/ml of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site.
  • Simplified Transcription/Translation of DNA Cloned in Plasmid or Lambda Vectors with a T7 Promoter
  • Express up to 500microg/ml of protein in one hour
  • Flexible detection of expressed proteins by Coomassie(R) staining or fluorescence or biotinylated labels
The E. coli S30 T7 High-Yield Protein Expression System is designed to express up to 500ug/ml of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site. The system provides an extract that contains T7 RNA polymerase for transcription and is deficient in OmpT endoproteinase and lon protease activity. All other necessary components in the system are optimized for translation. This results in greater stability and enhanced expression of target proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty