Would you like to log in again?
TNT T7 Quick Starter Bundle, Colorimetric, 1 each, Promega
Supplier: Promega
L1215
PAL1215EA 1121.5 USD InStock
PAL1215
TNT T7 Quick Starter Bundle, Colorimetric, 1 each, Promega
Protein Expression Protein Expression Systems
Purchase this bundle, and get the TNT T7 Quick Coupled Transcription/Translation System, your choice of Transcend Translation Detection System, and receive two cell-free expression-qualified expression vectors, pTNT and pCMVTNT Vectors, at no extra cost.
  • Starter Pack for Protein Expression, Detection and Interactions Analysis
  • TnT(R) T7 Quick Coupled Transcription/Translation System, a simple one-hour, one-tube reaction
  • Transcend™ Translation Detection Systems detects proteins with either chemiluminescent or colorimetric techniques
  • pTnT™ and pCMVTnT™ Vectors work to enhance cell-free expression
Get the tools you need in one bundle to start cell-free expression and detection of your proteins of interest. Use the system for in vitro analysis of protein:protein or protein:nucleic acid interactions, or simply verify the ability of your clone to express protein. Purchase this special bundle, and get the popular TNT T7 Quick Coupled Transcription/Translation System, your choice of Transcend Translation Detection System, and receive two cell-free expression-qualified expression vectors, pTNT and pCMVTNT Vectors, at no extra cost.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty