Would you like to log in again?
Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions, Promega
Supplier: Promega
L4960
PAL4960EA 287.34 USD InStock
PAL4960
Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions, Promega
Protein Extraction Protein Extraction Kits
Rabbit Reticulocyte Lysate Translation Systems are used in identification of mRNA species, characterization of their protein products and investigation of transcriptional and translational control.
  • In Vitro Translation of mRNA and Characterization of mRNA Products
  • Optimized for translation and ready to use
  • Contains an energy-regenerating system, a mix of tRNAs, hemin, potassium chloride and magnesium acetate
Rabbit Reticulocyte Lysate Translation Systems are utilized in the identification of mRNA species, the characterization of their protein products and the investigation of transcriptional and translational control. Rabbit Reticulocyte Lysate is prepared from New Zealand white rabbits using a standard protocol that ensures reliable and consistent reticulocyte production in each lot. After the reticulocytes are lysed, the extract is treated with micrococcal nuclease to destroy endogenous mRNA and thus reduce background translation to a minimum. The lysate contains the cellular components necessary for protein synthesis (tRNA, ribosomes, amino acids, initiation, elongation and termination factors).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty