Would you like to log in again?
TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions, Promega
Supplier: Promega
L5030
PAL5030EA 770.79 USD InStock
PAL5030
TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions, Promega
Protein Extraction Protein Extraction Kits
The TNT Coupled Wheat Germ Extract Systems are one-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.
  • Single-Tube Coupled Transcription/Translation System
  • Produces 2- to 6-fold more protein than standard wheat germ systems
  • Produce protein from linearized DNA in 1.5 hours
  • You choose the promoter
The TNT Coupled Wheat Germ Extract Systems offer researchers an alternative for eukaryotic cell-free protein expression: a one-tube, coupled transcription/translation system. The TNT Extract Systems greatly simplify the process and reduce the time required to obtain in vitro translation results. Standard wheat germ extract translations commonly use RNA synthesized in vitro from SP6 or T7 RNA polymerase promoters. This entire process requires separate reactions with several steps between each reaction. The TNT Extracts bypass many of these steps by incorporating transcription directly in the translation mix. Additionally, the TNT Extract reactions often produce significantly more protein (two- to sixfold) in a 1.5-hour reaction than do standard in vitro wheat germ extract translations using RNA templates. Magnesium Acetate, 25mM, and Potassium Chloride, 2.5M, can be used to optimize in vitro translation reactions in the TNT T7 Quick Coupled Transcription/Translation System, Flexi Rabbit Reticulocyte Lysate System and TNT Coupled Wheat Germ Extract System.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty