Would you like to log in again?
TNT T7 Quick for PCR DNA, 40 reactions, Promega
Supplier: Promega
L5540
PAL5540EA 874.24 USD InStock
PAL5540
TNT T7 Quick for PCR DNA, 40 reactions, Promega
Protein Expression Protein Expression Systems
TNT T7 Quick for PCR DNA is a rapid, convenient, coupled transcription/translation system designed for optimum protein expression from PCR templates.
  • Coupled Transcription/Translation System for Optimum Protein Expression from PCR Templates
  • Express protein directly from PCR, no cleanup needed
  • Up to 5 times more protein than standard translation reactions with linear templates
  • Includes Recombinant RNasin(R) Ribonuclease Inhibitor
TNT T7 Quick for PCR DNA is a rapid, convenient, coupled transcription/translation system designed for optimum protein expression from PCR templates. For most PCR templates, the TNT T7 Quick for PCR DNA reactions produce up to 5 times more protein than other commercially available kits. The PCR-generated DNA can be used directly from the amplification reaction or purified by numerous commercially available kits and traditional methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty