Would you like to log in again?
Exonuclease III
Supplier: Promega
M1811 M1815
PAM1811EA 138.25 USD InStock
PAM1811 PAM1815
Exonuclease III
Enzymes
Exonuclease III is a 3' --> 5' exonuclease specific for dsDNA.
  • For labeling in vitro, sequencing, mapping, mutagenesis,  duplex shortening
  • A 3´→5´ Exonuclease Specific for Double-Stranded DNA
  • Control deletion rate by varying incubation temperature
  • Inactivate Exonuclease III by heating at 75°C for 10 minutes
  • Supplied at a concentration of 150-200u/microl
Exonuclease III is a 3'-->5' exonuclease specific for double-stranded DNA. The enzyme catalyzes the stepwise removal of mononucleotides starting from a 3'-OH at nicks, blunt ends, recessed ends and 3'-overhangs of less than 4 bases, yielding nucleoside 5'-phosphates. Exonuclease III will also degrade DNA from 3'-phosphate ends due to its intrinsic 3'-phosphatase activity. In addition, the enzyme has apurinic endonuclease activity and ribonuclease H activity. Exonuclease III is used in conjunction with S1 nuclease for unidirectional deletion of sequences from the termini of DNA fragments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty