Would you like to log in again?
Recombinant Terminal Deoxynucleotidyl Transferase
Supplier: Promega
M1871 M1875
PAM1875EA 436.93 USD InStock
PAM1875 PAM1871
Recombinant Terminal Deoxynucleotidyl Transferase
Enzymes
TdT catalyzes the addition of mononucleotides to the terminal 3'-OH of a DNA initiator accompanied by the release of inorganic phosphate.
  • For labeling in vitro, and cDNA cloning
  • Adds Mononucleotides to the Terminal 3´-OH of ssDNA and Releases PPi
  • Adds mononucleotide tails to any type of 3´ end (3´ and 5´ overhangs or blunt ends) due to the presence of 1mM CoCl2 in the reaction buffer
  • Quality tested for labeling of apoptotic DNA ends using the procedure outlined in the DeadEnd™ Fluorometric TUNEL System Technical Bulletin #TB235 for each lot of enzyme
  • Supplied at a concentration of 30u/microl
Terminal Deoxynucleotidyl Transferase, Recombinant, catalyzes the repetitive addition of mononucleotides to the terminal 3'-OH of a DNA initiator accompanied by the release of inorganic phosphate. Single-stranded DNA is preferred as an initiator. Polymerization is not template-dependent. The addition of 1mM Co(2+) (as CoCl(2)) in the reaction buffer allows the tailing of 3'-ends with varying degrees of efficiency.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty