Would you like to log in again?
M-MLV Reverse Transcriptase, RNase H Minus, Point Mutant, Promega
Supplier: Promega
M3683 M3681 M3682
PAM3683EA 821.76 USD InStock
PAM3683 PAM3681 PAM3682
M-MLV Reverse Transcriptase, RNase H Minus, Point Mutant, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Point mutation of M-MLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.
  • High Yields of cDNA from Long RNA Templates (>5kb)
Moloney Murine Leukemia Virus Reverse Transcriptase, RNase H Minus (M-MLV RT [H-]), Point Mutant, is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long RNA templates (>5kb). The lack of RNase H activity is beneficial for this application, as RNase H can start to degrade templates when incubation times are long, as they may be when synthesizing long cDNAs. Although many researchers are successful in using M-MLV RT (H+) for analytical and some preparative cDNA applications, reverse transcriptases lacking RNase H activity provide another option to prepare long cDNAs and libraries containing a high percentage of full-length cDNA.
Caution: Storage temperature -30°C to -10°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty