Would you like to log in again?
GoTaq Long PCR Master Mix, 100 reactions, Promega
Supplier: Promega
GoTaq®
M4021
PAM4021EA 367.76 USD InStock
PAM4021
GoTaq Long PCR Master Mix, 100 reactions, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq Long PCR Master Mix contains the high-performance GoTaq Hot Start Polymerase in a specially formulated mixture with a proprietary thermal stable proofreading polymerase.
  • Efficiently Amplify Long DNA Amplicons
  • Optimized blend of thermostable DNA polymerases with enhanced processivity and proofreading
  • Hot-start master mix for convenient room temperature set-up
  • Optimized components for enhanced yield, sensitivity and specificity
GoTaq Long PCR Master Mix contains the high-performance GoTaq Hot Start Polymerase in a specially formulated mixture with a proprietary thermostable proofreading polymerase. This optimized enzyme mixture allows efficient amplification of up to 40kb from lambda DNA or 30kb from human genomic DNA. The presence of a proofreading enzyme to repair DNA mismatches and a highly processive polymerase allows the polymerase to continue to elongate the DNA much further, resulting in longer DNA amplification. The optimized formulation of the GoTaq Long PCR Master Mix components enables simple reaction setup and provides consistently efficient, accurate and robust amplification of long DNA amplicons.
The optimized formulation of the GoTaq® Long PCR Master Mix components enables simple reaction setup and provides consistently efficient, accurate, and robust amplification of long DNA amplicons.
A proofreading enzyme that repairs DNA mismatches and a highly processive polymerase allow the polymerase to continue to elongate the DNA further for longer DNA amplification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty