Would you like to log in again?
RNase ONE™ Ribonuclease
Supplier: Promega
M4261 M4265
PAM4261EA 173.75 USD InStock
PAM4261 PAM4265
RNase ONE™ Ribonuclease
Enzymes
Single-strand-specific RNA endonuclease. Cleaves between all ribonucleotides.
  • For sequencing and mapping
  • Degrades RNA to Cyclic Nucleotide Monophosphate (NMP) Intermediates
  • Ability to cleave a phosphodiester bond between any two ribonucleotides
  • Supplied at a concentration of 5-10u/microl
RNase ONE Ribonuclease is a 27kDa periplasmic enzyme from E. coli that catalyzes the degradation of RNA to cyclic nucleotide monophosphate (NMP) intermediates. Slower hydrolysis further catalyzes the degradation of these intermediates to 3'-NMPs. RNase ONE Ribonuclease is one of the few known RNases that can cleave a phosphodiester bond between any two ribonucleotides. RNase ONE Ribonuclease may be used to remove RNA from DNA preparations, for RNase protection assays and for mapping or quantitation of RNA by selective cleavage of single-stranded regions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty