Would you like to log in again?
AMV Reverse Transcriptase
Supplier: Promega
M5101 M5108
PAM5108EA 479.38 USD InStock
PAM5108 PAM5101
AMV Reverse Transcriptase
Enzymes
AMV RT catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids.
  • For sequencing, mutagenesis, cDNA synthesis, cDNA cloning and primer extension
  • Efficient Reverse Transcription of Templates with High Secondary Structure
  • DNA polymerization using template DNA, RNA or RNA:DNA hybrids
  • Requires a primer as well as magnesium or manganese
  • Stable at higher reaction temperatures (37-58°C)
  • Supplied at a concentration of 10u/microl (Cat.# M5101, M5108) or 20-25u/µl (Cat.# M9004)
Avian Myeloblastosis Virus Reverse Transcriptase (AMV RT) catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids. It requires a primer (DNA primers are more efficient than RNA primers) as well as Mg(2+) or Mn(2+). The enzyme possesses an intrinsic RNase H activity. Both nonionic detergents and sulfhydryl compounds stabilize the enzyme activity in vitro.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty