Would you like to log in again?
M-MLV Reverse Transcriptase Buffer Pack, Promega
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M5313
PAM5313EA 30.81 USD InStock
PAM5313
M-MLV Reverse Transcriptase Buffer Pack, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
M-MLV RT is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb).
  • RNA-Dependent DNA Polymerase for cDNA Synthesis
  • Enzyme supplied at a concentration of 200u/microl
Moloney Murine Leukemia Virus Reverse Transcriptase (M-MLV RT) is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb). The enzyme is a product of the pol gene of M-MLV and consists of a single subunit with a molecular weight of 71kDa. The RNase H activity of M-MLV RT is weaker than that of the commonly used Avian Myeloblastosis Virus (AMV) reverse transcriptase.
Caution: Storage temperature -30°C to -10°C
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty