Would you like to log in again?
RQ1 RNase-Free DNase
Supplier: Promega
M6101
PAM6101EA 80.21 USD InStock
PAM6101
RQ1 RNase-Free DNase
Enzymes
RQ1 (RNA Qualified) RNase-Free DNase is a DNase I that degrades both double-stranded and signle-stranded DNA.
  • For Mapping and transcription in vitro
  • Degrades Single-Stranded or Double-Stranded DNA
  • Produces 3´ hydroxyl oligonucleotides during degradation
  • Preparation qualified for use in applications where maintaining the integrity of RNA is critical
  • Supplied at a concentration of 1u/microl
RQ1 RNase-Free DNase is a preparation of deoxyribonuclease I that degrades single-stranded or double-stranded DNA to produce 3'-hydroxyl oligonucleotides. This preparation is qualified for use in applications where maintaining the integrity of RNA is critical.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty