Would you like to log in again?
GoTaq® G2 DNA Polymerase, Promega
Supplier: Promega
GoTaq®
M7848 M7841 M7845
PAM7848EA 420.2 USD InStock
PAM7848 PAM7845 PAM7841
GoTaq® G2 DNA Polymerase, Promega
Nucleic Acid Reagents End-point PCR Enzymes and Kits
GoTaq G2 DNA Polymerase is supplied with 5X Green GoTaq Reaction Buffer and 5X Colorless GoTaq Reaction Buffer.
  • Next-Generation GoTaq(R) for Robust, Reliable PCR Amplification
  • Full-length Taq DNA polymerase that exhibits 5´→3´ exonuclease activity
  • Direct-to-gel convenience
  • 5X Reaction Buffer with MgCl2 to simplify reaction setup
  • Supplied at a concentration of 5u/microl
The second generation of GoTaq products, GoTaq G2 DNA Polymerase reliably amplifies a wide range of PCR templates and provides high-performing results due to improved manufacturing processes, increased reliability and consistency. GoTaq G2 DNA Polymerase is supplied with 5X Green GoTaq Reaction Buffer and 5X Colorless GoTaq Reaction Buffer. Both buffers contain MgCl2 at a concentration of 7.5mM for a final concentration of 1.5mM in the 1X reaction. GoTaq G2 DNA Polymerase is supplied in a proprietary formulation containing 50% glycerol with buffers designed for enhanced amplification. The enzyme is a full-length form of Taq DNA polymerase that exhibits 5'-->3' exonuclease activity. The 5X Green GoTaq Reaction Buffer contains two dyes (blue and yellow) that separate during electrophoresis to indicate migration progress. The colorless buffer is used when direct fluorescence or absorbance readings are required without prior purification of the amplified DNA from the PCR.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty