Would you like to log in again?
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega
N1110 N1120 N1150 N1130
PAN1110EA 209.21 USD InStock
PAN1110 PAN1120 PAN1130 PAN1150
Nano-Glo® Luciferase Assay System, Promega, Promega
Assays Cellular Assays
The Nano-Glo Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media.
  • Add-Mix-Measure assay for NanoLuc(R) Luciferase
  • Bright NanoLuc(R) reporter enables use in applications where sensitivity is limited
  • Add-and-read simplicity allows scaling from bench to HTS
  • Extended bright light output optimized for batch and continuous processing
The Nano-Glo Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media. The reagent is prepared by mixing Nano-Glo Luciferase Assay Substrate and Nano-Glo Luciferase Assay Buffer. The reagent contains an integral lysis buffer allowing use directly on cells expressing NanoLuc luciferase or the culture media when luciferase is secreted. Nano-Glo Luciferase Assay Reagent is a dedicated product for the detection of NanoLuc Luciferase. For more details on NanoLuc Luciferase, visit the NanoLuc Luciferase Technology page.
Nano-Glo Luciferase Assay Reagent is a dedicated product for the detection of NanoLuc Luciferase.
The reagent contains an integral lysis buffer that can be used directly on cells expressing NanoLuc luciferase or the culture media when luciferase is secreted.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty