Would you like to log in again?
NanoBRET Nano-Glo Substrate, Promega
Supplier: Promega
N1573 N1571 N1572
PAN1573EA 11762.73 USD InStock
PAN1573 PAN1572 PAN1571
NanoBRET Nano-Glo Substrate, Promega
Assays Cellular Assays
The NanoBRET™ Nano-Glo(R) Substrate is the substrate used by NanoLuc(R) Luciferase to generate the donor signal in several NanoBRET™ Assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty