Would you like to log in again?
pGEM-7Zf(+) Vector, 20 µg, Promega
Supplier: Promega
P2251
PAP2251EA 168.59 USD InStock
PAP2251
pGEM-7Zf(+) Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.
  • Standard Cloning Vectors and Templates for In vitro Transcription and Circular ssDNA Production
  • Blue/white screening for easily identifying recombinant clones
  • In vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Orientation of the f1 origin differentiates vectors (+ and -)
The pGEM-7Zf(+) and pGEM-7Zf(-) Vectors are derivatives of the pGEM-3Zf(+) Vector and contain the origin of replication of the filamentous phage f1. These plasmids serve as standard cloning vectors, as templates for in vitro transcription and can be used for the production of circular ssDNA. These plasmids contain SP6 and T7 RNA polymerase promoters flanking a region of multiple cloning sites within the alpha-peptide coding region of beta-galactosidase. Insertional inactivation of the alpha-peptide allows recombinant clones to be identified directly by color screening on indicator plates when using appropriate E. coli strains (e.g., JM109). The multiple cloning region is unique and includes restriction sites for ApaI, AatII, SphI, XbaI, XhoI, EcoRI, KpnI, SmaI, ClaI, HindIII, BamHI, SacI, BstXI and NsiI. This arrangement is designed specifically for generating unidirectional deletions with the Erase-a-Base System. pGEM-7Zf(+) and pGEM-7Zf(-) Vectors are identical except for the orientation of the f1 origin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty