Would you like to log in again?
EcoR I (Blue/White Cloning Qualified), Restriction Enzymes, Promega
Supplier: Promega
R4014
PAR4014EA 156.13 USD InStock
PAR4014
EcoR I (Blue/White Cloning Qualified), Restriction Enzymes, Promega
Nucleic Acid Reagents Restriction Enzymes
G/AATT C C TTAA\G
  • Capable of digesting DNA in 15 minutes or less
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Available at high concentration, containing 25,000 and 50,000 units of EcoRI at a concentration of 40-80u/microl
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty