Would you like to log in again?
SacI, Restriction Enzymes, Promega
Supplier: Promega
R6065 R6061
PAR6065EA 334.2 USD InStock
PAR6065 PAR6061
SacI, Restriction Enzymes, Promega
Nucleic Acid Reagents Restriction Enzymes
G AGCT/C C\TCGA G
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty