Would you like to log in again?
SmaI
Supplier: Promega
R6121 R6125
PAR6121EA 129.03 USD InStock
PAR6121 PAR6125
SmaI
Nucleic Acid Reagents Restriction Enzymes
CCC/GGG GGG\CCC
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
  • Available at high concentration, containing 5,000 units of SmaI at a concentration of 40-80u/microl
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty