Would you like to log in again?
PvuI
Supplier: Promega
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R6321 R6325
PAR6321EA 103.24 USD InStock
PAR6321 PAR6325
PvuI
Nucleic Acid Reagents Restriction Enzymes
CG AT/CG GC\TA GC
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty